ENA402267324 Enamine

sodium 3-cyclohexyl-3-oxoprop-1-en-1-olate

  • Cas Number : 1344821-52-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131388

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131388
Availability details for
Quantity
  • Availability
  • Add to Cart