ENA402267325 Enamine

5-fluoro-N,N,3-trimethyl-1-benzofuran-2-carboxamide

  • MDL number MFCD01762404
  • Cas Number : 81718-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131390

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131390
Availability details for
Quantity
  • Availability
  • Add to Cart