ENA402267326 Enamine

methyl 3-[(4-cyanophenyl)sulfanyl]propanoate

  • MDL number MFCD12563644
  • Cas Number : 1154018-52-6
  • Molecular Weight : 221.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-131392

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131392
Availability details for
Quantity
  • Availability
  • Add to Cart