ENA402267329 Enamine

2-(2,5-dioxoimidazolidin-1-yl)acetonitrile

  • MDL number MFCD09949178
  • Cas Number : 1016787-64-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131396

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131396
Availability details for
Quantity
  • Availability
  • Add to Cart