ENA402267334 Enamine

4-(3-bromopropyl)pyridine hydrobromide

  • MDL number MFCD20484231
  • Cas Number : 64262-18-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126...128
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131426

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131426
Availability details for
Quantity
  • Availability
  • Add to Cart