ENA402267336 Enamine

4-(1H-1,2,3-triazol-1-yl)butan-1-amine

  • MDL number MFCD21997101
  • Cas Number : 325490-78-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131428

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131428
Availability details for
Quantity
  • Availability
  • Add to Cart