ENA402267346 Enamine

(5-chloro-1,3-thiazol-2-yl)methanol

  • MDL number MFCD12405637
  • Cas Number : 50398-78-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128711

Safety Information

signalword Warning
pcodes P305+P351+P338, P337+P313,P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P312,P330,P332+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-128711
pictograms GHS07
hstatements H302,H315,H320,H335
Availability details for
Quantity
  • Availability
  • Add to Cart