ENA402267351 Enamine

3-(pyrrolidin-1-yl)pyridine-4-carboxylic acid

  • MDL number MFCD03411731
  • Cas Number : 1257901-68-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131467

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131467
Availability details for
Quantity
  • Availability
  • Add to Cart