ENA402267355 Enamine

4-chloro-8-fluoro-2-methyl-3-(propan-2-yl)quinoline

  • MDL number MFCD17246879
  • Cas Number : 1343392-03-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 47...49
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131471

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131471
Availability details for
Quantity
  • Availability
  • Add to Cart