ENA402267357 Enamine

tert-butyl N-(5-carbamoyl-4-methyl-1,3-thiazol-2-yl)carbamate

  • MDL number MFCD25371721
  • Cas Number : 1461707-68-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131473

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131473
Availability details for
Quantity
  • Availability
  • Add to Cart