ENA402267358 Enamine

5-acetamidopyridine-2-carboxylic acid

  • MDL number MFCD15475043
  • Cas Number : 86873-62-3
  • Molecular Weight : 180.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131475

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131475
Availability details for
Quantity
  • Availability
  • Add to Cart