ENA402267363 Enamine

3-hydrazinyl-4,5-dihydro-1,2,4-triazin-5-one

  • MDL number MFCD01325250
  • Cas Number : 21383-22-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104373
Availability details for
Quantity
  • Availability
  • Add to Cart