ENA402267364 Enamine

1-(2,6-difluorophenyl)-1H-1,3-benzodiazole-2-thiol

  • MDL number MFCD11543137
  • Cas Number : 1038333-41-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 255...257
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131485

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131485
Availability details for
Quantity
  • Availability
  • Add to Cart