ENA402267366 Enamine

(dicyclopropyl-4H-1,2,4-triazol-3-yl)methanol

  • MDL number MFCD24197541
  • Cas Number : 1461707-62-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160...162
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131487

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131487
Availability details for
Quantity
  • Availability
  • Add to Cart