ENA402267372 Enamine

4-chloro-3-(methylsulfanyl)quinoline

  • MDL number MFCD20270592
  • Cas Number : 83936-07-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131502

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131502
Availability details for
Quantity
  • Availability
  • Add to Cart