ENA402267375 Enamine

(2S)-2-(phenylformamido)butanoic acid

  • MDL number MFCD24141807
  • Cas Number : 87068-75-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108...110
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131540

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131540
Availability details for
Quantity
  • Availability
  • Add to Cart