ENA402267376 Enamine

4-bromo-N-(2,2-dimethoxyethyl)-1-methyl-1H-pyrrole-2-carboxamide

  • MDL number MFCD14587426
  • Cas Number : 1249514-57-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131545

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131545
Availability details for
Quantity
  • Availability
  • Add to Cart