ENA402267378 Enamine

(4-sulfanylphenyl)methanol

  • MDL number MFCD00041529
  • Cas Number : 53339-53-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54...56
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131552

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-131552
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart