ENA402267380 Enamine

ethyl 3-formyl-2,2-dimethylcyclopropane-1-carboxylate

  • MDL number MFCD11500113
  • Cas Number : 66692-75-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131568

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131568
Availability details for
Quantity
  • Availability
  • Add to Cart