ENA402267381 Enamine

2-ethoxypropane-1,3-diamine

  • MDL number MFCD17285605
  • Cas Number : 210232-59-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131579

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131579
Availability details for
Quantity
  • Availability
  • Add to Cart