ENA402267382 Enamine

4-(but-3-yn-1-yloxy)benzoic acid

  • MDL number MFCD09939454
  • Cas Number : 1017024-01-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131583

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131583
Availability details for
Quantity
  • Availability
  • Add to Cart