ENA402267384 Enamine

1-(2-cyclopropylpyridin-4-yl)methanamine

  • MDL number MFCD18255581
  • Cas Number : 852877-61-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 85
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131585

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131585
Availability details for
Quantity
  • Availability
  • Add to Cart