ENA402267385 Enamine

ethyl 3-{[(benzyloxy)carbonyl]amino}propanoate

  • MDL number MFCD16242554
  • Cas Number : 161617-96-5
  • Molecular Weight : 251.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 3-{[(benzyloxy)carbonyl]amino}propanoate
Inchi Code 1S/C13H17NO4/c1-2-17-12(15)8-9-14-13(16)18-10-11-6-4-3-5-7-11/h3-7H,2,8-10H2,1H3,(H,14,16)
Inchi Key IFXFHOYJVRMTTO-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-131586

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131586
Availability details for
Quantity
  • Availability
  • Add to Cart