ENA402267388 Enamine

2-chloro-6-[2-(trifluoromethoxy)phenyl]pyridine

  • MDL number MFCD18403109
  • Cas Number : 1261856-69-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131589

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131589
Availability details for
Quantity
  • Availability
  • Add to Cart