ENA402267395 Enamine

2-chloro-1-(chloromethyl)-4-nitrobenzene

  • MDL number MFCD12025222
  • Cas Number : 50274-95-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 50...52
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131601

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131601
Availability details for
Quantity
  • Availability
  • Add to Cart