ENA402267397 Enamine

methyl 3-(chlorosulfonyl)-5-fluorobenzoate

  • MDL number MFCD12169474
  • Cas Number : 1155909-75-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131603
Availability details for
Quantity
  • Availability
  • Add to Cart