ENA402267400 Enamine

(5-methylthiophen-3-yl)boronic acid

  • MDL number MFCD18250237
  • Cas Number : 930303-82-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160...162
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131606

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131606
Availability details for
Quantity
  • Availability
  • Add to Cart