ENA402267407 Enamine

5-hydroxy-3-methyl-1H-pyrazole-4-carbonitrile

  • MDL number MFCD25371726

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131613

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131613
Availability details for
Quantity
  • Availability
  • Add to Cart