ENA402267417 Enamine

4-chloro-1-(trimethylsilyl)pent-1-yn-3-one

  • MDL number MFCD09031633
  • Cas Number : 914358-91-1
  • Molecular Weight : 188.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-131656

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131656
Availability details for
Quantity
  • Availability
  • Add to Cart