ENA402267418 Enamine

1-(3-chloropropoxy)-2,3-dimethylbenzene

  • MDL number MFCD09731154
  • Cas Number : 31264-52-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131661

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131661
Availability details for
Quantity
  • Availability
  • Add to Cart