ENA402267422 Enamine

N,N-diethyl-3-(piperidin-4-yl)propanamide

  • MDL number MFCD16693121
  • Cas Number : 1249844-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Country of Origin Ukraine

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128924
Availability details for
Quantity
  • Availability
  • Add to Cart