ENA402267423 Enamine

5-(thiophen-3-yl)-10,13-dioxa-4,6-diazatricyclo[7.4.0.0^{3,7}]trideca-1(9),2,4,7-tetraene

  • MDL number MFCD11547590
  • Cas Number : 1097819-60-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 279...281
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131676

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131676
Availability details for
Quantity
  • Availability
  • Add to Cart