ENA402267424 Enamine

methyl 2-[2-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetate

  • MDL number MFCD12913983
  • Cas Number : 956229-86-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65...67
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131677

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131677
Availability details for
Quantity
  • Availability
  • Add to Cart