ENA402267425 Enamine

2-cyclopropylpyridine-4-carboxylic acid

  • MDL number MFCD13187883
  • Cas Number : 1216171-07-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 285-287
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131679

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131679
Availability details for
Quantity
  • Availability
  • Add to Cart