ENA402267429 Enamine

[4-(2,6-difluorophenyl)-1,3-thiazol-2-yl]methanol

  • MDL number MFCD17251268
  • Cas Number : 1342657-77-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131686

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131686
Availability details for
Quantity
  • Availability
  • Add to Cart