ENA402267432 Enamine

2-(1,2,4-oxadiazol-3-yl)morpholine hydrochloride

  • MDL number MFCD25371733
  • Cas Number : 1461708-58-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131690

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131690
Availability details for
Quantity
  • Availability
  • Add to Cart