ENA402267438 Enamine

1-(1-cyclopropylethyl)-5-methyl-1H-pyrazol-3-amine

  • MDL number MFCD16845359
  • Cas Number : 1248970-88-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129000

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129000
Availability details for
Quantity
  • Availability
  • Add to Cart