ENA402267443 Enamine

methyl 2-(5-methyl-4-nitro-1H-pyrazol-1-yl)acetate

  • MDL number MFCD18374178
  • Cas Number : 1245772-53-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95...97
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131764

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131764
Availability details for
Quantity
  • Availability
  • Add to Cart