ENA402267447 Enamine

diethyl[3-(piperidin-4-yl)propyl]amine

  • MDL number MFCD16693184
  • Cas Number : 127425-06-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131771

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131771
Availability details for
Quantity
  • Availability
  • Add to Cart