ENA402267448 Enamine

3-{2-bromoimidazo[2,1-b][1,3,4]thiadiazol-6-yl}pyridine

  • MDL number MFCD11209938
  • Cas Number : 1094279-90-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 230...232
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131773

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131773
Availability details for
Quantity
  • Availability
  • Add to Cart