ENA402267452 Enamine

tert-butyl N-(5-cyano-4-methyl-1,3-thiazol-2-yl)carbamate

  • MDL number MFCD25371737
  • Cas Number : 1461708-14-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148...150
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131777

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131777
Availability details for
Quantity
  • Availability
  • Add to Cart