ENA402267453 Enamine

4-(3-cyclopropyl-1,2,4-oxadiazol-5-yl)-2-methylaniline

  • MDL number MFCD11123462
  • Cas Number : 1036444-77-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 87...89
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131778

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131778
Availability details for
Quantity
  • Availability
  • Add to Cart