ENA402267463 Enamine

4-(azocan-1-yl)-3-fluoroaniline

  • MDL number MFCD10689275
  • Cas Number : 1092279-99-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131803

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131803
Availability details for
Quantity
  • Availability
  • Add to Cart