ENA402267464 Enamine

2-[methyl(3-methylbut-2-en-1-yl)amino]ethan-1-ol

  • MDL number MFCD16152168
  • Cas Number : 1250038-76-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131806

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131806
Availability details for
Quantity
  • Availability
  • Add to Cart