ENA402267471 Enamine

4-tert-butylpyrrolidin-2-one

  • MDL number MFCD22412986
  • Cas Number : 824966-98-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130...132
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129040

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129040
Availability details for
Quantity
  • Availability
  • Add to Cart