ENA402267474 Enamine

(3-chloro-5-methylphenyl)methanol

  • MDL number MFCD18393675
  • Cas Number : 116069-80-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-131822

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131822
Availability details for
Quantity
  • Availability
  • Add to Cart