ENA402267475 Enamine

benzyl 2-sulfamoylpropanoate

  • MDL number MFCD25371743
  • Cas Number : 1461705-34-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83...85
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131824

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131824
Availability details for
Quantity
  • Availability
  • Add to Cart