ENA402267480 Enamine

butyl 2-(4-cyanopiperidin-1-yl)acetate

  • MDL number MFCD20047817
  • Cas Number : 1457358-72-3
  • Molecular Weight : 224.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131849

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131849
Availability details for
Quantity
  • Availability
  • Add to Cart