ENA402267481 Enamine

ethyl 2-{[(naphthalen-1-yl)methyl]amino}-2,3-dihydro-1H-indene-2-carboxylate

  • MDL number MFCD25371745
  • Cas Number : 1385585-13-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-131850

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131850
Availability details for
Quantity
  • Availability
  • Add to Cart