ENA402267487 Enamine

2-(3,4-diethoxyphenyl)-2-methylpropanoic acid

  • MDL number MFCD17338838
  • Cas Number : 1271063-42-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 74...76
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131888

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131888
Availability details for
Quantity
  • Availability
  • Add to Cart